1H,1H,2H,2H-Heptadecafluorodecyl methacrylate ≥95% stabilized
Supplier: Matrix Scientific
Synonyms:
2-(Perfluorooctyl)ethyl methacrylate
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 225 ppm
002216-100G
002216-25G
101775-870EA
253.85
USD
101775-870
101775-868
1H,1H,2H,2H-Heptadecafluorodecyl methacrylate ≥95% stabilized
1H,1H,2H,2H-Heptadecafluorodecyl methacrylate
TCA listed: T
Hazard: Corrosive/Irritant/Refrigerate
Formula:
C₁₄H₉F₁₇O₂ MW: 532.2 g/mol Boiling Pt: 120 °C (4 mmHg) Density: 1.59 |
MDL Number:
MFCD00042307 CAS Number: 1996-88-9 |
Specification Test Results
Boiling point | 120°/4mm |
Density | 1.59 |
ND20 | 1.343 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...