5-Fluoro-2-methylbenzylamine

002165-1G
101775-824EA 193.64 USD
101775-824
5-Fluoro-2-methylbenzylamine
5-Fluoro-2-methylbenzylamine

Hazard: Corrosive/Air Sensitive

Formula: C₈H₁₀FN
MDL Number: MFCD01631526
CAS Number: 261951-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR