1-Chloro-2-iodotetrafluoroethane ≥97%

Supplier: Matrix Scientific

002139-5G
101775-808EA 475.38 USD
101775-808
1-Chloro-2-iodotetrafluoroethane ≥97%
1-Chloro-2-iodotetrafluoroethane

Hazard: Toxic/Light Sensitive

Formula: C₂ClF₄I
Boiling Pt: 56…57 °C
MDL Number: MFCD00069092
CAS Number: 421-78-3
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 56-57°
ND20 1.39

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR