3-Chloro-2-fluorophenylacetonitrile ≥98%
Supplier: Matrix Scientific
002114-25G
002114-5G
002114-1G
101775-794EA
800
USD
101775-794
101775-792
101775-790
3-Chloro-2-fluorophenylacetonitrile ≥98%
3-Chloro-2-fluorophenylacetonitrile
Hazard: Toxic
Formula:
C₈H₅ClFN MW: 169.5800 g/mol |
MDL Number:
MFCD01631552 CAS Number: 261762-98-5 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...