3-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde ≥95%
Supplier: Matrix Scientific
002041-5G
002041-25G
101775-702EA
63.25
USD
101775-702
101775-700
3-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde ≥95%
3-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde
Hazard: Toxic
Formula:
C₉H₆F₄O₂ Boiling Pt: 95…97 °C (9 mmHg) Density: 1.393 |
CAS Number:
35295-35-3 |
Specification Test Results
Boiling point | 95-97°/9mm |
Density | 1.393 |
ND20 | 1.454 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...