3-Fluoro-2-(trifluoromethyl)benzonitrile
Supplier: Matrix Scientific
002010-250MG
002010-1G
101775-674EA
89.23
USD
101775-674
101775-672
3-Fluoro-2-(trifluoromethyl)benzonitrile
3-Fluoro-2-(trifluoromethyl)benzonitrile
Hazard: Toxic/Irritant
Formula:
C₈H₃F₄N MW: 189.11 g/mol |
MDL Number:
MFCD01631566 CAS Number: 261951-81-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...