1-Chloro-1-fluoroethylene ≥97%

Supplier: Matrix Scientific

001914-25G
101775-598EA 976.92 USD
101775-598
1-Chloro-1-fluoroethylene ≥97%
1-Chloro-1-fluoroethylene

Hazard: Flammable Gas

Formula: C₂H₂ClF
Boiling Pt: –24 °C
Melting Pt: –169 °C
CAS Number: 2317-91-1

Order Now

Specification Test Results

Boiling point -24°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR