1-Chloro-1,2,2-trifluorocyclobutane ≥97%

Supplier: Matrix Scientific

001900-5G
101775-592EA 113.85 USD
101775-592
1-Chloro-1,2,2-trifluorocyclobutane ≥97%
1-Chloro-1,2,2-trifluorocyclobutane

Hazard: Flammable

Formula: C₄H₄ClF₃
Boiling Pt: 81 °C
Density: 1.34
MDL Number: MFCD00042139
CAS Number: 661-71-2

Order Now

Specification Test Results

Boiling point 81°
Density 1.34
ND20 1.358

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR