4-Bromo-2-trifluoromethylphenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

001870-5G 001870-1G
101775-568EA 140 USD
101775-568 101775-566
4-Bromo-2-trifluoromethylphenyl isothiocyanate ≥98%
4-Bromo-2-trifluoromethylphenyl isothiocyanate

Hazard: Toxic/Corrosive/Refrigerate/Lachrymator

Formula: C₈H₃BrF₃NS
Boiling Pt: 130 °C (7 mmHg)
Density: 1.67
MDL Number: MFCD00041086
CAS Number: 206559-46-8

Order Now

Specification Test Results

Boiling point 130°/7mm
Density 1.67
ND20 1.613

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR