2-Amino-4-(trifluoromethyl)thiophenol hydrochloride ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-(trifluoromethyl)benzenethiol hydrochloride

001789-5G
101775-552EA 272.31 USD
101775-552
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride ≥98%
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride

Hazard: Toxic

Formula: C₇H₇ClF₃NS
Melting Pt: 196…198 °C
MDL Number: MFCD00042600
CAS Number: 4274-38-8

Order Now

Specification Test Results

Melting point 196-198°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR