2,2,3,3,3-Pentafluoropropylamine ≥97%

Supplier: Matrix Scientific

001567-1G 001567-5G
101775-452EA 123.08 USD
101775-452 101775-450
2,2,3,3,3-Pentafluoropropylamine ≥97%
2,2,3,3,3-Pentafluoropropylamine

Hazard: Corrosive/Refrigerate

Formula: C₃H₄F₅N
Boiling Pt: 49…50 °C
Density: 1.4
MDL Number: MFCD00042459
CAS Number: 422-03-7

Order Now

Specification Test Results

Boiling point 49-50°
Density 1.4
ND20 1.297

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR