1H,1H,2H-Perfluoro-1-butene ≥97%

Supplier: Matrix Scientific

001565-5G
101775-448EA 561.54 USD
101775-448
1H,1H,2H-Perfluoro-1-butene ≥97%
1H,1H,2H-Perfluoro-1-butene

Hazard: Flammable Gas

Formula: C₄H₃F₅
MW: 146.06 g/mol
Boiling Pt: 3…6 °C
CAS Number: 374-27-6

Order Now

Specification Test Results

Boiling point 3-6°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR