1,1,3,3,3-Pentafluoropropene ≥97%

Supplier: Matrix Scientific

Synonyms: FC-1225zc

001551-25G 001551-100G
101775-424EA 2100 USD
101775-424 101775-422
1,1,3,3,3-Pentafluoropropene ≥97%
1,1,3,3,3-Pentafluoropropene

Hazard: Flammable Gas

Formula: C₃HF₅
MW: 132.03 g/mol
Boiling Pt: –21 °C
Melting Pt: –153 °C
CAS Number: 690-27-7

Order Now

Specification Test Results

Boiling point -21°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR