Perfluoro-3,5,5-trimethylhexanoic acid ≥84%

001441-5G 001441-25G
101775-384EA 116.85 USD
101775-384 101775-382
Perfluoro-3,5,5-trimethylhexanoic acid ≥84%
Perfluoro-3,5,5-trimethylhexanoic acid

Hazard: Corrosive

Formula: C₉HF₁₇O₂
Boiling Pt: 200 °C
MDL Number: MFCD01320752
CAS Number: 238403-51-5

Order Now

Specification Test Results

Boiling point 200°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR