Perfluoro-2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecanoyl fluoride ≥90%

Supplier: Matrix Scientific

001414-5G
101775-380EA 146.15 USD
101775-380
Perfluoro-2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecanoyl fluoride ≥90%
Perfluoro-2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecanoyl fluoride

Balance is other HFPO oligomer acid fluorides


TCA listed: T
Hazard: Corrosive

Formula: C₁₈F₃₆O₆
Boiling Pt: 231…235 °C
Density: 1.8
MDL Number: MFCD00054661
CAS Number: 13252-15-8

Order Now

Specification Test Results

Boiling point 231-235°
Density 1.8

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR