2,6-Dichloro-4-(trifluoromethyl)nicotinonitrile ≥98%

Supplier: Matrix Scientific

001302-5G 001302-1G
101775-342EA 116.92 USD
101775-342 101775-340
2,6-Dichloro-4-(trifluoromethyl)nicotinonitrile ≥98%
2,6-Dichloro-4-(trifluoromethyl)nicotinonitrile

Hazard: Toxic

Formula: C₇HCl₂F₃N₂
Melting Pt: 39…40 °C
MDL Number: MFCD00084977
CAS Number: 13600-42-5

Order Now

Specification Test Results

Melting point 35-37°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR