Phenol : chloroform : iso-amyl alcohol (125:24:1), OmniPur® (Citrate Buffered pH 4.5), Millipore®

Danger

Synonyms: Carbolic acid, Hydroxybenzene, Phenic acid

6800-100ML
CA6800
Phenol : chloroform : iso-amyl alcohol (125:24:1), OmniPur® (Citrate Buffered pH 4.5), Millipore®
Phenol : Chloroform : iso-Amyl alcohol
Density: 1.28
Storage Temperature: Refrigerator
MDL Number: MFCD00133763
CAS Number: 136112-00-0
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless liquid
Assay [phenol] 99.0% min
pH at 25°C 4.3-4.7
UV Absorbance at 330 nm 0.2AU max
UV Absorbance at 405 nm 0.1AU max
UV Absorbance at 510 nm 0.1AU max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR