Chloramphenicol, powder

Supplier: Corning

Synonyms: Chloromycetin®(R,R)-ChloramphenicolTega-Cetin

61-239-RI 61-239-RI
45000-618EA 125.92 USD
CA45000-618 45000-618
Chloramphenicol, powder
Chloramphenicol

Chloramphenicol is a common antibiotic which inhibits microbial protein synthesis by preventing peptide bond formation. It is effective against Gram-positive and Gram-negative bacteria.


Packaging: Shipping temperature: Ambient

Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078159
CAS Number: 56-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR