    Sort by:
Dimethyl allylphosphonate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Allylphosphonic acid dimethyl ester. Grade:tech. 85, Melting Point C. Boiling Point C:41-42*/0. 5mm. C5H11O3P. 757-54-0.