    Sort by:
  • Search Within Results
Magnesium 8-anilinonaphthalene-1-sulphonate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   [Hydrophobic fluorescent probe] CAS Number: 18108-68-4 MDL Number: MFCD00065353 Molecular Formula: C16H13NO3S Molecular Weight: 620.98 Purity/Ana...