Vitamin K1 (phytomenadione, phylloquinone) ≥97% USP
Synonyms:
2-Methyl-3-((7R,11R,E)-3,7,11,15-tetramethylhexadec-2-en-1-yl)naphthalene-1,4-dione
PH195-25GM
PH195-5GM
PH195-100GM
95034-938EA
3648.93
USD
95034-938
95034-946
95034-944
Vitamin K1 (phytomenadione, phylloquinone) ≥97% USP
Vitamin K1 (Phytomenadione, Phylloquinone)
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₃₁H₄₆O₂ MW: 450.6958 g/mol Density: 0.984 Storage Temperature: Ambient |
MDL Number:
MFCD00214063 CAS Number: 81818-54-4 EINECS: 201-564-2 |
Specification Test Results
Assay (%) | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...