Malt extract, liquid
Supplier: SPECTRUM CHEMICAL MFG CORP
M3217-500GM
700012-948EA
299
USD
700012-948
Malt extract, liquid
Malt extract
Malt Extract, Liquid is used in cosmetic face masks and toning lotions as a nutrient and texturizer.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD00131570 CAS Number: 8002-48-0 EINECS: 232-310-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...