Riboflavine-5'-phosphate sodium salt dihydrate USP
Synonyms:
Sodium (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,3,4-trihydroxypentyl hydrogenphosphate dihydrate
RI106-100GM
RI106-1KGBL
RI106-25GM
700009-340EA
217.91
USD
700009-340
700009-336
700009-338
Riboflavine-5'-phosphate sodium salt dihydrate USP
Riboflavine-5'-phosphate sodium salt dihydrate
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₁₇H₂₀N₄NaO₉P·2H₂O MW: 514.36 g/mol (478.34 anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00065362 CAS Number: 6184-17-4 EINECS: 204-988-6 |
Specification Test Results
Assay (%) | 73 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...