Guanosine ≥98%, Reagent Grade
Synonyms:
2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
GU105-25GM
GU105-5GM
GU105-100GM
GU105-500GM
700009-140EA
72.2
USD
700009-140
700009-132
700009-136
700009-138
Guanosine ≥98%, Reagent Grade
Guanosine
Formula:
C₁₀H₁₃N₅O₅ MW: 283.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010182 CAS Number: 118-00-3 EINECS: 204-227-8 |
Specification Test Results
Assay (%) | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...