DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)
Synonyms:
DNA sodium salt
, Thymonucleic acid sodium salt
DE120-100GM
DE120-25GM
DE120-1KGBL
700009-120EA
204.48
USD
700009-120
700009-116
700009-118
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)
DNA sodium salt (Deoxyribonucleic acid sodium salt)
Deoxyribonucleic Acid, Sodium Salt is a nucleic acid composed of three macromolecules important for all forms of life. The molecule encodes the genetic map used in functioning living viruses and organisms.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD00130921 CAS Number: 100403-24-5 EINECS: 309-566-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...