Ectoine ≥95%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic acid

E3002-1GM E3002-10GM
700008-938EA 998 USD
700008-938 700008-940
Ectoine ≥95%
Ectoine

Ectoine, also known as 1,4,5,6-tetrahydro-2-methyl-4-pyrimidinecarboxylic acid, is a natural compound used as an ingredient in sun protection and skin care products which stabilizes proteins and other cellular structures against UV irradiation and dryness.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₀N₂O₂
MW: 142.1558 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03419286
CAS Number: 96702-03-3
EINECS: 431-910-1

Order Now

Specification Test Results

Assay (%) 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR