(±)-2-Pyrrolidinone-5-carboxylic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: DL-2-Pyrrolidone-5-carboxylic acidDL-Pyroglutamic acid5-Oxopyrrolidine-2-carboxylic acid

P2757-500GM
700008-876EA 437.04 USD
700008-876
(±)-2-Pyrrolidinone-5-carboxylic acid
(±)-2-Pyrrolidinone-5-carboxylic acid

DL-Pyroglutamic Acid is also known as Pidolic acid or 5-Oxo-proline. It is an amino acid used on the skin to retain moisture and as a dietary supplement.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅H₇NO₃
MW: 129.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064322
CAS Number: 149-87-1
EINECS: 205-748-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR