Cetyldimethylethylammonium bromide ≥99%, high purity

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Ethylhexadecyldimethylammonium BromideEthylhexadecyldimethylammonium bromideEthyl(1-hexadecyl)dimethylammonium bromideDimethylethylhexadecylammonium bromide

E1054-100GM E1054-2.5KG E1054-500GM E1054-12KGBL E1054-5GM
700007-620EA 2034 USD
700007-620 700007-618 700007-616 700007-622 700007-624
Cetyldimethylethylammonium bromide ≥99%, high purity
Cetyldimethylethylammonium bromide

Ethylhexadecyldimethylammonium Bromide, High Purity is a quaternary ammonium compound (QAC) commonly used as a cationic surfactant.


The High Purity grade denotes that this chemical is the highest quality commercially available. Spectrum Chemical manufactured High Purity grade products meet the toughest regulatory standards for quality and purity.

Formula: C₂₀H₄₄BrN
MW: 378.4741 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011774
CAS Number: 124-03-8
EINECS: 204-672-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay (%) 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR