Dimethyldioctadecylammonium chloride (DODMAC) ≥97%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Dimethyldistearylammonium ChlorideDimethyldi-n-octadecylammonium chlorideDimethyldioctadecylammonium chloride

D1496-100GM
700007-598EA 109 USD
700007-598
Dimethyldioctadecylammonium chloride (DODMAC) ≥97%
Dimethyldioctadecylammonium chloride (DODMAC)
Formula: C₃₈H₈₀ClN
MW: 586.5015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00055540
CAS Number: 107-64-2
EINECS: 203-508-2

Order Now

Specification Test Results

Assay (%) 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR