DL-Carnitine hydrochloride ≥99%
Synonyms:
3-Hydroxy-4-(trimethylammonio)butanoate hydrochloride
CA197-100GM
CA197-25GM
700007-584EA
67.4
USD
700007-584
700007-582
DL-Carnitine hydrochloride ≥99%
DL-Carnitine hydrochloride
DL-Carnitine Hydrochloride is a quaternary ammonium compound bio-synthesized from the amino acids lysine and methionine. It used in cosmetic and personal care products as a humectant and skin conditioning agent. It appears as a solid that is easily soluble in cold water, soluble in methanol and insoluble in diethyl ether and acetone.
Formula:
C₇H₁₆ClNO₃ MW: 197.66 Storage Temperature: Ambient |
MDL Number:
MFCD00011904 CAS Number: 461-05-2 EINECS: 207-309-1 |
Specification Test Results
Assay (%) | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...