(-)-Carveol (mixture of cis- and trans-isomers) ≥97%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
p-Mentha-6,8-dien-2-ol
, (5R)-5-Isopropenyl-2-methyl-2-cyclohexenol
C2750-50GM
C2750-10GM
700003-126EA
770
USD
700003-126
700003-124
(-)-Carveol (mixture of cis- and trans-isomers) ≥97%
(-)-Carveol (mixture of cis- and trans-isomers)
(-)-Carveol is a colorless liquid that is soluble in oil but not water with a scent similar to spearmint and caraway. Because of its odor, it is often used in cosmetics and fragrances and in the food industry as a flavor additive.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₀H₁₆O MW: 152.24 g/mol Density: 1.49 Storage Temperature: Ambient |
MDL Number:
MFCD00869995 CAS Number: 99-48-9 EINECS: 202-757-4 |
Specification Test Results
Assay (%) | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...