Would you like to log in again?
RSA™ Inserts, Precision Point, 200 uL, MicroSolv
Supplier: MicroSolv Technology Corp
9502S-02ND-RS
These inserts are available today for HPLC, GC, LCMS & GCMS
These inserts are best used when working with low volumes of biological or basic compounds that might interact or adsorb to borosilicate glass. Also available in silanized, deactivated for critical biological applications. When RSA glass is silanized using our process, a virtually complete chemically inert vial is produced.
Starting with the highest quality TYPE 1, 33 expansion pharmaceutical grade glass, these inserts are made with the unique RSA process. The result is borosilicate glass with extremely low amount of surface metals, virtually zero silanols, and no potential pH effects due to the glass.
Ordering information: Supplied with attached plastic spring; available in packs of 100.
ORDER
Expand All / Collapse All
Description Capacity Dimensions Supplier No. VWR Catalog Number Unit Price Quantity
Inserts, with attached plastic springs, silanized, use with 12x32mm vials 300 µL 6x29 mm 9502S-02ND-RS 97051-398 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty