2-Undecenoic acid ≥70.0%
Supplier: TCI America
U0032-5G
TCU0032-005GEA
140.02
USD
TCU0032-005G
2-Undecenoic acid ≥70.0%
2-Undecenoic acid
CAS Number: 4189-02-0
MDL Number: MFCD00040909
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Freezing point (°C): 11
Formula:
C₁₁H₂₀O₂ MW: 184.28 g/mol |
MDL Number:
MFCD00040909 CAS Number: 4189-02-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...