Triethylene glycol monoisopropyl ether ≥98.0%

Supplier: TCI America

Synonyms: Isopropyl Triglycol

T1373-5G
TCT1373-005GEA 201.89 USD
TCT1373-005G
Triethylene glycol monoisopropyl ether ≥98.0%
Triethylene glycol monoisopropyl ether

CAS Number: 29681-21-8
MDL Number: MFCD00145451
Molecular Formula: C9H20O4
Molecular Weight: 192.26
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 125
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Isopropyl Triglycol

Formula: C₉H₂₀O₄
MW: 192.26 g/mol
Boiling Pt: 125 °C
Density: 1.00 (20 °C)
MDL Number: MFCD00145451
CAS Number: 29681-21-8
https://us.vwr.com/stibo/web/std.lang.all/53/99/28995399.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR