4-Methyl-2-nitroanisole ≥98.0%

Supplier: TCI America
Warning

Synonyms: 4-Methoxy-3-nitrotoluene4-Methyl-2-nitroanisole4-Methoxy-4-nitrotoluene1-Methoxy-4-methyl-2-nitrobenzeneMethyl 2-nitro-p-tolyl ether

M1205-25G
TCM1205-025GEA 40.19 USD
TCM1205-025G
4-Methyl-2-nitroanisole ≥98.0%
4-Methyl-2-nitroanisole

CAS Number: 119-10-8
MDL Number: MFCD00024540
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 274
Flash Point (°C): 113
Freezing point (°C): 8
Specific Gravity (20/20): 1.21


Other Product Names / Synonyms: 1-Methoxy-4-methyl-2-nitrobenzene, 4-Methyl-2-nitroanisole

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 274 °C
Density: 1.21 (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00024540
CAS Number: 119-10-8
EINECS: 204-296-4
UN: 2446
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/49/73/28974973.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR