3,3-Diphenylpropionic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3,3-Diphenylpropanoic acid

D1779-25G
TCD1779-025GEA 322 USD
TCD1779-025G
3,3-Diphenylpropionic acid ≥97.0% (by titrimetric analysis)
3,3-Diphenylpropionic acid

CAS Number: 606-83-7
MDL Number: MFCD00002717
Molecular Formula: C15H14O2
Molecular Weight: 226.28
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 153
Flash Point (°C): 113

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 153 °C
Flash Pt: 113 °C
MDL Number: MFCD00002717
CAS Number: 606-83-7
EINECS: 210-125-4
https://us.vwr.com/stibo/web/std.lang.all/85/53/28978553.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR