1,3-Diisopropylthiourea ≥99.0% (by total nitrogen basis)
Supplier: TCI America
D0253-25G
TCD0253-025GEA
73.08
USD
TCD0253-025G
1,3-Diisopropylthiourea ≥99.0% (by total nitrogen basis)
1,3-Diisopropylthiourea
CAS Number: 2986-17-6
MDL Number: MFCD00040485
Molecular Formula: C7H16N2S
Molecular Weight: 160.28
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 144
Formula:
C₇H₁₆N₂S Melting Pt: 144 °C |
MDL Number:
MFCD00040485 CAS Number: 2986-17-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...