TCIAB1016

B1016-5G
TCB1016-005GEA 287.22 USD
TCB1016-005G
TCIAB1016
CAS Number: 15430-62-3
MDL Number: MFCD00040509
Molecular Formula: C5H7BrN2O
Molecular Weight: 191.03
Purity/Analysis Method: 90.0% (GC)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 57

Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: N,N-Dimethyl-2-bromo-2-cyanoacetamide, 2-(N,N-Dimethylcarbamoyl)-2-bromoacetonitrile
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR