2-Bromo-n-octanoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B0627-25G
TCB0627-025GEA 153.2 USD
TCB0627-025G
2-Bromo-n-octanoic acid ≥97.0% (by GC, titration analysis)
2-Bromo-n-octanoic acid

CAS Number: 2623-82-7
MDL Number: MFCD00004219
Molecular Formula: C8H15BrO2
Molecular Weight: 223.11
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 164
Flash Point (°C): 110
Specific Gravity (20/20): 1.26

Formula: C₈H₁₅BrO₂
MW: 223.11 g/mol
Boiling Pt: 164 °C
Density: 1.26 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004219
CAS Number: 2623-82-7
https://us.vwr.com/stibo/web/std.lang.all/25/58/28932558.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR