Brommequat bromide ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Bromocholine bromide2-Bromoethyltrimethylammonium bromide(2-Bromoethyl)trimethylammonium bromideBromocholine Bromide

B0577-25G
TCB0577-025GEA 84.65 USD
TCB0577-025G
Brommequat bromide ≥99.0% (by titrimetric analysis)
Brommequat bromide

CAS Number: 2758-06-7
MDL Number: MFCD00011865
Molecular Formula: C5H13Br2N
Molecular Weight: 246.97
Purity/Analysis Method: 99.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2-Bromoethyltrimethylammonium Bromide

Formula: BrCH₂CH₂N(CH₃)₃Br
MW: 246.97 g/mol
MDL Number: MFCD00011865
CAS Number: 2758-06-7
EINECS: 220-419-4
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/25/18/28932518.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR