1-Bromo-2-butene ≥83.0%

Supplier: TCI America

Synonyms: 1-Bromo-2-buteneCrotyl bromide

B0562-25ML
TCB0562-025MLEA 197.48 USD
TCB0562-025ML
1-Bromo-2-butene ≥83.0%
1-Bromo-2-butene

CAS Number: 4784-77-4
MDL Number: MFCD00000248
Molecular Formula: C4H7Br
Molecular Weight: 135.00
Purity/Analysis Method: 83.0% (GC)
Form: Clear Liquid
Boiling point (°C): 107
Flash Point (°C): 13
Specific Gravity (20/20): 1.34
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Bromo-2-butene

Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 107 °C
Density: 1.34 (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00000248
CAS Number: 4784-77-4
EINECS: 225-332-5
https://us.vwr.com/stibo/web/std.lang.all/25/05/28932505.jpg

Order Now

Specification Test Results

Purity/Analysis Method 83.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR