Sodium-4-amino-1,5-naphthalenedisulfonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Monosodium 4-Amino-1,5-naphthalenedisulphonate
, 1-Naphthylamine-4,8-disulphonic acid Monosodium Salt
, Monosodium 4-Amino-1,5-naphthalenedisulfonate
, 4-Amino-1,5-naphthalenedisulfonic acid Monosodium Salt
, 1-Naphthylamine-4,8-disulfonic acid Monosodium Salt
, 4-Amino-1,5-naphthalenedisulphonic acid Monosodium Salt
CAS Number: 85328-80-9
MDL Number: MFCD00021519
Molecular Formula: C10H9NO6S2
Molecular Weight: 325.28
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 1-Naphthylamine-4,8-disulfonic Acid Monosodium Salt, 4-Amino-1,5-naphthalenedisulfonic Acid Monosodium Salt, Monosodium 4-Amino-1,5-naphthalenedisulfonate
Formula:
C₁₀H₉NO₆S₂ MW: 325.28 g/mol |
MDL Number:
MFCD00021519 CAS Number: 85328-80-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...