Bis(4-nitrophenyl) disulfide ≥97.0%

Supplier: TCI America

Synonyms: 4-Nitrophenyl disulphide1,2-Bis(4-nitrophenyl)disulfane4,4'-Dinitrodiphenyl Disulphide4-Nitrophenyl disulfide1,2-Bis(4-nitrophenyl)disulphane4,4'-Dinitrodiphenyl Disulfide

B0504-25G
TCB0504-025GEA 58.87 USD
TCB0504-025G
Bis(4-nitrophenyl) disulfide ≥97.0%
Bis(4-nitrophenyl) disulphide

CAS Number: 100-32-3
MDL Number: MFCD00003573
Molecular Formula: C12H8N2O4S2
Molecular Weight: 308.33
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 183


Product Categories: Synthetic Organic Chemistry, Sulfenylation
Other Product Names / Synonyms: 4,4'-Dinitrodiphenyl Disulfide, 4-Nitrophenyl Disulfide

Formula: C₁₂H₈N₂O₄S₂
MW: 308.34 g/mol
Melting Pt: 183 °C
MDL Number: MFCD00003573
CAS Number: 100-32-3
EINECS: 202-840-5
https://us.vwr.com/stibo/web/std.lang.all/24/58/28932458.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR