N,N-Bis(2-hydroxyethyl)-3-chloraniline ≥90.0%

Supplier: TCI America

Synonyms: 2-[(3-Chlorophenyl)(2-hydroxyethyl)amino]ethanol

B0490-25G
TCB0490-025GEA 56 USD
TCB0490-025G
N,N-Bis(2-hydroxyethyl)-3-chloraniline ≥90.0%
N,N-Bis(2-hydroxyethyl)-3-chloraniline

CAS Number: 92-00-2
MDL Number: MFCD00002852
Molecular Formula: C10H14ClNO2
Molecular Weight: 215.68
Purity/Analysis Method: 90.0% (GC)
Form: Crystal
Melting point (°C): 84


Other Product Names / Synonyms: 3-Chloro-N,N-di(2-hydroxyethyl)aniline, N-(3-Chlorophenyl)diethanolamine, 2,2'-[(3-Chlorophenyl)imino]diethanol, N,N-Di(2-hydroxyethyl)-3-chloroaniline

Formula: C₁₀H₁₄ClNO₂
Melting Pt: 84 °C
MDL Number: MFCD00002852
CAS Number: 92-00-2
EINECS: 202-115-3
https://us.vwr.com/stibo/web/std.lang.all/24/47/28932447.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR