Bis(2,4-dinitrophenyl)disulfide ≥80.0% (by HPLC)
Synonyms:
2,2',4,4'-Tetranitrodiphenyl Disulphide
, 2,2',4,4'-Tetranitrodiphenyl Disulfide
B0487-25G
TCB0487-025GEA
173.16
USD
TCB0487-025G
Bis(2,4-dinitrophenyl)disulfide ≥80.0% (by HPLC)
Bis(2,4-dinitrophenyl)disulfide
CAS Number: 2217-55-2
MDL Number: MFCD00059178
Molecular Formula: C12H6N4O8S2
Molecular Weight: 398.32
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 2,2',4,4'-Tetranitrodiphenyl Disulfide
Formula:
C₁₂H₆N₄O₈S₂ MW: 398.32 g/mol |
MDL Number:
MFCD00059178 CAS Number: 2217-55-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...