N-Benzyl-o-phenetidine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
N-Benzyl-2-ethoxyaniline
B0431-25G
TCB0431-025GEA
362
USD
TCB0431-025G
N-Benzyl-o-phenetidine ≥98.0% (by titrimetric analysis)
N-Benzyl-o-phenetidine
CAS Number: 13371-95-4
MDL Number: MFCD00059373
Molecular Formula: C15H17NO
Molecular Weight: 227.31
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 32
Other Product Names / Synonyms: N-Benzyl-2-ethoxyaniline
Formula:
C₁₅H₁₇NO MW: 227.31 g/mol Melting Pt: 32 °C |
MDL Number:
MFCD00059373 CAS Number: 13371-95-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...