N-Benzylacetamide ≥98.0%
Supplier: TCI America
Synonyms:
N-Acetylbenzylamine
B0404-500G
B0404-25G
TCB0404-500GEA
676.81
USD
TCB0404-500G
TCB0404-025G
N-Benzylacetamide ≥98.0%
N-Benzylacetamide
CAS Number: 588-46-5
MDL Number: MFCD00059204
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 157
Melting point (°C): 61
Other Product Names / Synonyms: N-Acetylbenzylamine
Formula:
C₉H₁₁NO MW: 149.19 g/mol Boiling Pt: 157 °C Melting Pt: 61 °C |
MDL Number:
MFCD00059204 CAS Number: 588-46-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...