TCIAB0369

B0369-100MG
TCB0369-100MGEA 107.21 USD
TCB0369-100MG
TCIAB0369
CAS Number: 106629-90-7
MDL Number: MFCD00060036
Molecular Formula: C46H44Cl2N8O6
Molecular Weight: 875.81
Form: Crystal
Color: Pale Yellow
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR