2-(2-Benzothiazolyl)-3,5-diphenyltetrazolium bromide ≥95.0% (by titrimetric analysis)

Supplier: TCI America

B0333-100MG
TCB0333-100MGEA 76.09 USD
TCB0333-100MG
2-(2-Benzothiazolyl)-3,5-diphenyltetrazolium bromide ≥95.0% (by titrimetric analysis)
2-(2-Benzothiazolyl)-3,5-diphenyltetrazolium bromide

CAS Number: 55699-64-4
MDL Number: MFCD00135111
Molecular Formula: C20H14BrN5S
Molecular Weight: 436.33
Purity/Analysis Method: 95.0% (T)
Form: Crystal

Formula: C₂₀H₁₄BrN₅S
MDL Number: MFCD00135111
CAS Number: 55699-64-4
https://us.vwr.com/stibo/web/std.lang.all/23/59/28932359.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR