Benzoyl-DL-phenylalanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Bz-DL-Phe-OH

B0302-10G
TCB0302-010GEA 208 USD
TCB0302-010G
Benzoyl-DL-phenylalanine ≥98.0% (by titrimetric analysis)
Benzoyl-DL-phenylalanine

CAS Number: 2901-76-0
MDL Number: MFCD00037252
Molecular Formula: C16H15NO3
Molecular Weight: 269.30
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 187

Formula: C₁₆H₁₅NO₃
MW: 269.30 g/mol
Melting Pt: 187 °C
MDL Number: MFCD00037252
CAS Number: 2901-76-0
https://us.vwr.com/stibo/web/std.lang.all/23/45/28932345.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR